Assay Details
Target Gene Details
Entrez Gene ID: | 80796 |
Gene Name: | calmodulin 4 |
Gene Aliases: |
2310037J09Rik, AI649108, DD112, Scarf |
Location: |
Chr.13:3837757-3838671 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Calm4 | NM_020036.4 | 1 | 601 | NP_064420.2 |
| AI131917.1 | 1 | 359 | ||
| AK003648.1 | 1 | 602 | BAB22914.1 | |
| AK009664.1 | 1 | 604 | BAB26425.1 | |
| AK009956.1 | 1 | 602 | BAB26608.1 | |
| AY293058.1 | 1 | 584 | AAQ56119.1 | |
| BC060284.1 | 1 | 587 | AAH60284.1 | |
| BC145379.1 | 1 | 557 |
More Information
Set Membership: |
|

Genomic Map