Assay Details
Target Gene Details
Entrez Gene ID: | 238205 |
Gene Name: | leucine rich repeat and fibronectin type III domain containing 5 |
Gene Aliases: |
AI427653, AI604817, C130061B21, mKIAA4208 |
Location: |
Chr.12:61522817-61857930 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lrfn5 | NM_178714.4 | 3 | 1078 | NP_848829.2 |
AK034245.1 | 3 | 1079 | BAC28645.1 | |
AK048443.1 | 3 | 1164 | BAC33339.1 | |
AK083920.1 | 3 | 2272 | ||
AK220546.1 | 3 | 1762 | BAD90535.1 | |
BC052038.1 | 3 | 908 | AAH52038.1 |
