Assay Details
Target Gene Details
Entrez Gene ID: | 109700 |
Gene Name: | integrin alpha 1 |
Gene Aliases: |
CD49A, E130012M19Rik, Vla1 |
Location: |
Chr.13:114958079-115101964 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itga1 | NM_001033228.3 | 14 | 2125 | NP_001028400.2 |
| AK035870.1 | 14 | 2126 | BAE20498.1 | |
| AK040830.1 | 5 | 549 | ||
| AK043769.1 | 14 | 2064 | BAE20643.1 | |
| AK137169.1 | 14 | 1890 | ||
| BC014765.1 | 14 | 1989 |
More Information
Set Membership: |
|

Genomic Map