Assay Details
Target Gene Details
Entrez Gene ID: | 13162 |
Gene Name: | solute carrier family 6 (neurotransmitter transporter, dopamine), member 3 |
Gene Aliases: |
DAT, Dat1 |
Location: |
Chr.13:73535528-73578672 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc6a3 | NM_010020.3 | 2 | 330 | NP_034150.1 |
| AF109072.1 | 2 | 253 | AAD19643.1 | |
| AF109391.1 | 1 | 178 | AAF85795.1 | |
| AJ238309.1 | 1 | 186 | CAB51926.1 | |
| AK032629.1 | 2 | 288 | BAC27959.1 | |
| AK136885.1 | 2 | 275 | BAE23154.1 | |
| BC054119.1 | 2 | 330 | AAH54119.1 | |
| U16265.1 | AAC52283.1 |
More Information
Set Membership: |
|

Genomic Map