Assay Details
Target Gene Details
Entrez Gene ID: | 17129 |
Gene Name: | SMAD family member 5 |
Gene Aliases: |
1110051M15Rik, AI451355, Dwf-C, Madh5, MusMLP |
Location: |
Chr.13:56702988-56742379 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smad5 | NM_001164041.1 | 3 | 1261 | NP_001157513.1 |
| NM_001164042.1 | 3 | 881 | NP_001157514.1 | |
| NM_008541.3 | 3 | 858 | NP_032567.1 | |
| AF063006.1 | 3 | 1262 | AAC83580.1 | |
| AK018077.1 | 3 | 830 | BAB31063.1 | |
| AK082997.1 | 3 | 882 | BAC38724.1 | |
| AK139765.1 | 3 | 803 | ||
| BC048233.1 | AAH48233.1 | |||
| BC050001.1 | 3 | 761 | AAH50001.2 | |
| U58993.1 | 3 | 837 | AAB07871.1 | |
| U77638.1 | 3 | 1007 | AAB39737.1 |
More Information
Set Membership: |
|

Genomic Map