Assay Details
Target Gene Details
Entrez Gene ID: | 67252 |
Gene Name: | CAP, adenylate cyclase-associated protein, 2 (yeast) |
Gene Aliases: |
2810452G09Rik, AV045235 |
Location: |
Chr.13:46501903-46650281 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cap2 | NM_026056.4 | 13 | 2655 | NP_080332.1 |
| AK033071.1 | 13 | 2411 | BAC28143.1 | |
| AK049813.1 | 10 | 2098 | ||
| AK076737.1 | 9 | 2075 |
More Information
Set Membership: |
|

Genomic Map