Assay Details
Target Gene Details
Entrez Gene ID: | 74724 |
Gene Name: | RIKEN cDNA 4930512B01 gene |
Gene Aliases: |
4930472D12Rik |
Location: |
Chr.12:69790344-69841880 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930512B01Rik | NR_033573.1 | |||
| AK015558.1 |
Target Gene Details
Entrez Gene ID: | 399510 |
Gene Name: | mitogen-activated protein kinase kinase kinase kinase 5 |
Gene Aliases: |
4432415E19Rik, GCKR, KHS, MAPKKKK5 |
Location: |
Chr.12:69803757-69893163 on Build GRCm38 |
Assay Gene Location: | Within Exon 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Map4k5 | NM_201519.2 | 32 | 3128 | NP_958927.2 |
| AK019468.1 | 32 | 2918 | BAB31739.1 | |
| AK032123.1 | 32 | 3014 | ||
| AK053775.1 | 32 | 3129 | BAC35517.1 | |
| AK080965.1 | 8 | 2929 | ||
| AK084891.1 | 32 | 2836 | BAC39305.1 | |
| AK198812.1 | 1 | 250 | ||
| BC002309.1 | 13 | 1182 | AAH02309.1 | |
| BC040381.1 | 31 | 2977 | AAH40381.2 | |
| BC048173.1 | 31 | 3010 | ||
| BC057930.1 | 16 | 1402 | AAH57930.1 |
More Information
Set Membership: |
|

Genomic Map