Assay Details
Target Gene Details
Entrez Gene ID: | 218194 |
Gene Name: | phosphatase and actin regulator 1 |
Gene Aliases: |
9630030F18Rik, Rpel1 |
Location: |
Chr.13:42680571-43138526 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Phactr1 | AK047654.1 | |||
AY993932.1 | AAY42814.1 | |||
BY130551.1 | 1 | 210 |
