Assay Details
Target Gene Details
Entrez Gene ID: | 18676 |
Gene Name: | PHD finger protein 2 |
Gene Aliases: |
GRC5 |
Location: |
Chr.13:48801750-48870885 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Phf2 | NM_011078.3 | 18 | 2645 | NP_035208.2 |
AB221590.1 | 7 | 1031 | ||
AF043726.1 | 18 | 2645 | AAD21792.1 | |
AK134358.1 | 4 | 473 | BAE22113.1 | |
AK172995.1 | 16 | 2352 | BAD32273.1 | |
BC051633.1 | 18 | 2643 | AAH51633.1 |
