Assay Details
Target Gene Details
Entrez Gene ID: | 12522 |
Gene Name: | CD83 antigen |
Gene Aliases: |
- |
Location: |
Chr.13:43785112-43803133 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cd83 | NM_001289915.1 | 5 | 1869 | NP_001276844.1 |
NM_009856.3 | 5 | 1876 | NP_033986.1 | |
AJ245551.1 | 5 | 1787 | CAB63843.1 | |
AK075888.1 | 5 | 1806 | BAC36032.1 | |
AK139410.1 | 5 | 1804 | BAE23999.1 | |
AK154862.1 | 5 | 1804 | BAE32886.1 | |
AK156386.1 | 5 | 1805 | BAE33694.1 | |
AK172396.1 | 5 | 1792 | BAE42984.1 | |
AK172449.1 | 5 | 1805 | BAE43011.1 | |
AK172466.1 | 5 | 1799 | BAE43020.1 | |
AK172515.1 | 4 | 1899 | BAE43044.1 | |
AV359852.1 |
