Assay Details
Target Gene Details
Entrez Gene ID: | 27015 |
Gene Name: | polymerase (DNA directed), kappa |
Gene Aliases: |
Dinb1 |
Location: |
Chr.13:96480689-96542648 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Polk | NM_012048.2 | 15 | 3971 | NP_036178.1 |
| AF163571.1 | 15 | 3971 | AAF02541.1 | |
| AK014087.1 | 3 | 1630 | BAB29148.1 | |
| BC052820.1 | 12 | 3233 | AAH52820.1 |
More Information
Set Membership: |
|

Genomic Map