Assay Details
Target Gene Details
Entrez Gene ID: | 105513 |
Gene Name: | charged multivesicular body protein 7 |
Gene Aliases: |
4930596K11Rik, 6330407G04Rik, AI450338, AW550775 |
Location: |
Chr.14:69716979-69732570 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chmp7 | NM_134078.4 | 10 | 2492 | NP_598839.2 |
| AK011732.1 | 1 | 873 | BAC25343.1 | |
| AK031882.1 | 1 | 793 | BAC27592.1 | |
| AK179878.1 | 1 | 134 | ||
| AK182423.1 | 1 | 134 | ||
| AK190385.1 | 1 | 134 | ||
| AK191247.1 | 1 | 134 | ||
| AK196156.1 | 1 | 134 | ||
| BC024115.1 | 10 | 2480 | AAH24115.1 | |
| BC033365.1 | 8 | 2922 | AAH33365.1 |
More Information
Set Membership: |
|

Genomic Map