Assay Details
Target Gene Details
Entrez Gene ID: | 238988 |
Gene Name: | ELKS/RAB6-interacting/CAST family member 2 |
Gene Aliases: |
6430531D06, CAST, CAST1/ERC2, D14Ertd171e, ELKS2alpha |
Location: |
Chr.14:27622410-28478537 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Erc2 | NM_177814.4 | 3 | 1310 | NP_808482.2 |
| AK032385.1 | 5 | 1334 | BAC27848.1 | |
| AK147213.1 | 4 | 1349 | BAE27770.1 | |
| AK162818.1 | 3 | 441 | BAE37067.1 | |
| AY356530.1 | 2 | 997 | AAR14793.1 | |
| AY356531.1 | 2 | 997 | AAR14794.1 | |
| AY356532.1 | 2 | 997 | AAR14795.1 | |
| AY356533.1 | 2 | 997 | AAR14796.1 | |
| BC056760.1 | 3 | 1310 | AAH56760.1 |
More Information
Set Membership: |
|

Genomic Map