Assay Details
Target Gene Details
Entrez Gene ID: | 108800 |
Gene Name: | stonin 2 |
Gene Aliases: |
4933401N24Rik, AI425999, AW544599 |
Location: |
Chr.12:91633009-91786436 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ston2 | NM_175367.6 | 5 | 2597 | NP_780576.1 |
| AK036612.1 | 5 | 2598 | BAC29507.1 | |
| BC132531.1 | 4 | 2183 | ||
| BC138054.1 | 4 | 2183 |
More Information
Set Membership: |
|

Genomic Map