Assay Details
Target Gene Details
Entrez Gene ID: | 72125 |
Gene Name: | APC membrane recruitment 2 |
Gene Aliases: |
2600011E07Rik, Fam123a |
Location: |
Chr.14:60373293-60388197 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Amer2 | NM_001164705.1 | NP_001158177.1 | ||
| NM_028113.3 | 1 | 941 | NP_082389.1 | |
| AK011185.1 | BAB27452.1 | |||
| AK032651.1 | 1 | 882 | BAC27972.1 | |
| AK139177.1 | 1 | 888 | BAE23912.1 | |
| BC056350.1 | AAH56350.1 |

Genomic Map