Assay Details
Target Gene Details
Entrez Gene ID: | 238880 |
Gene Name: | actin, beta-like 2 |
Gene Aliases: |
4732495G21Rik |
Location: |
Chr.13:111255013-111257749 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Actbl2 | NM_175497.3 | 1 | 1363 | NP_780706.1 |
| AK029110.1 | 1 | 1364 | BAC26303.1 | |
| AK029130.1 | 1 | 1361 | BAC26313.1 | |
| BC111904.1 | 1 | 1329 | ||
| BC112429.1 | 1 | 1329 |
More Information
Set Membership: |
|

Genomic Map