Assay Details
Target Gene Details
Entrez Gene ID: | 18109 |
Gene Name: | v-myc myelocytomatosis viral related oncogene, neuroblastoma derived (avian) |
Gene Aliases: |
N-myc, Nmyc, Nmyc-1, Nmyc1, bHLHe37, c-nmyc |
Location: |
Chr.12:12936093-12941836 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mycn | NM_008709.3 | 2 | 633 | NP_032735.3 |
| AK144126.1 | 2 | 633 | ||
| AK146696.1 | 2 | 634 | BAE27366.1 | |
| AK146913.1 | 2 | 631 | BAE27525.1 | |
| BC005453.1 | 2 | 517 | AAH05453.3 | |
| BC049783.1 | 1 | 562 | AAH49783.1 |
More Information
Set Membership: |
|

Genomic Map