Assay Details
Target Gene Details
Entrez Gene ID: | 57266 |
Gene Name: | chemokine (C-X-C motif) ligand 14 |
Gene Aliases: |
1110031L23Rik, 1200006I23Rik, AI414372, BMAC, BRAK, KS1, Kec, MIP-2g, MIP2gamma, NJAC, Scyb14, bolekine |
Location: |
Chr.13:56288643-56296792 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cxcl14 | NM_019568.2 | 4 | 1445 | NP_062514.2 |
| AB041614.1 | 4 | 1445 | BAA95097.1 | |
| AF144754.1 | 4 | 1269 | AAF66694.1 | |
| AF192557.1 | 4 | 1443 | AAF03753.1 | |
| AK004008.1 | 1 | 332 | ||
| AK004615.1 | 4 | 1289 | BAB23411.1 | |
| AK076112.1 | 4 | 1287 | BAC36192.1 | |
| BC079661.1 | 3 | 1085 | AAH79661.1 | |
| BC147274.1 | ||||
| BC147275.1 |
More Information
Set Membership: |
|

Genomic Map