Assay Details
Target Gene Details
Entrez Gene ID: | 14776 |
Gene Name: | glutathione peroxidase 2 |
Gene Aliases: |
GI-GPx, GPx-GI, GSHPx-2, GSHPx-GI |
Location: |
Chr.12:76792335-76795554 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpx2 | NM_030677.2 | 1 | 66 | NP_109602.2 |
| BC039658.1 | AAH39658.1 | |||
| CF612186.1 | 1 | 95 |
More Information
Set Membership: |
|

Genomic Map