Assay Details
Target Gene Details
Entrez Gene ID: | 72873 |
Gene Name: | coiled-coil domain containing 176 |
Gene Aliases: |
2900006K08Rik, AI427784 |
Location: |
Chr.12:84409068-84441763 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc176 | NM_028377.3 | NP_082653.2 | ||
| AK133381.1 | BAE21625.1 | |||
| BC151099.1 | ||||
| BC151103.1 |
Target Gene Details
Entrez Gene ID: | 66381 |
Gene Name: | ring finger protein 113A2 |
Gene Aliases: |
2310020H19Rik, AI426758, Rnf113a |
Location: |
Chr.12:84417200-84418578 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnf113a2 | NM_025525.2 | 1 | 1138 | NP_079801.2 |
| AK009420.1 | 1 | 1159 | BAB26275.1 | |
| AK016795.1 | 1 | 1098 | ||
| BC116426.2 | 1 | 1079 |
More Information
Set Membership: |
|

Genomic Map