Assay Details
Target Gene Details
Entrez Gene ID: | 238896 |
Gene Name: | cell division cycle 20B |
Gene Aliases: |
EG238896, EG622422 |
Location: |
Chr.13:113032954-113121019 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdc20b | NM_001281487.1 | NP_001268416.1 |
Target Gene Details
Entrez Gene ID: | 69590 |
Gene Name: | glutathione peroxidase 8 (putative) |
Gene Aliases: |
2310016C16Rik, AU017063 |
Location: |
Chr.13:113042763-113046388 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpx8 | NM_027127.2 | 3 | 569 | NP_081403.1 |
| AK009378.1 | 3 | 570 | BAB26254.1 | |
| BC019664.1 | 3 | 568 | AAH19664.1 |
More Information
Set Membership: |
|

Genomic Map