Assay Details
Target Gene Details
Entrez Gene ID: | 78038 |
Gene Name: | methylcrotonoyl-Coenzyme A carboxylase 2 (beta) |
Gene Aliases: |
4930552N12Rik, MCCB |
Location: |
Chr.13:99948532-100015639 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mccc2 | NM_030026.2 | 17 | 1777 | NP_084302.1 |
| AK132265.1 | 17 | 1757 | BAE21068.1 | |
| AK143233.1 | 17 | 1778 | BAE25319.1 | |
| AK145564.1 | 17 | 1729 | BAE26513.1 | |
| AK146844.1 | 17 | 1737 | BAE27475.1 | |
| AK146865.1 | 17 | 1727 | BAE27489.1 | |
| BC027640.1 | AAH27640.1 | |||
| BC058987.1 | 9 | 791 | AAH58987.1 | |
| BC141193.1 | 17 | 1743 | ||
| BC141194.1 | 17 | 1743 |
More Information
Set Membership: |
|

Genomic Map