Assay Details
Target Gene Details
Entrez Gene ID: | 56196 |
Gene Name: | tyrosyl-DNA phosphodiesterase 2 |
Gene Aliases: |
D13Ertd656e, Ttrap |
Location: |
Chr.13:24831647-24842153 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tdp2 | NM_019551.2 | 7 | 1260 | NP_062424.1 |
| AJ251328.1 | 7 | 1234 | CAB92971.1 | |
| AK009089.1 | ||||
| AK135925.1 | 7 | 1272 | ||
| AK167707.1 | 6 | 1056 | BAE39751.1 | |
| BC132511.1 | 7 | 1223 | ||
| BC132513.1 | 7 | 1223 |

Genomic Map