Assay Details
Target Gene Details
Entrez Gene ID: | 105501 |
Gene Name: | abhydrolase domain containing 4 |
Gene Aliases: |
1110035H23Rik, AI429574, Abh4 |
Location: |
Chr.14:54254129-54269169 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abhd4 | NM_001205181.1 | 3 | 740 | NP_001192110.1 |
| NM_134076.2 | 3 | 485 | NP_598837.2 | |
| AK030250.1 | 3 | 681 | BAC26863.1 | |
| AK049366.1 | 3 | 466 | BAC33711.1 | |
| AK152595.1 | 3 | 486 | BAE31344.1 | |
| AK161853.1 | 3 | 435 | BAE36606.1 | |
| AK164655.1 | 3 | 445 | ||
| BC003982.1 | 1 | 125 | AAH03982.1 | |
| BC017532.1 | 3 | 420 | AAH17532.1 |

Genomic Map