Assay Details
Target Gene Details
Entrez Gene ID: | 238875 |
Gene Name: | Grb2-binding adaptor, transmembrane |
Gene Aliases: |
9830130M13Rik |
Location: |
Chr.13:110352614-110357273 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gapt | NM_177713.3 | 3 | 483 | NP_808381.1 |
| AK036534.1 | 3 | 484 | BAC29466.1 | |
| AK155882.1 | 3 | 491 | BAE33480.1 | |
| BC116629.1 | 1 | 330 | ||
| BC118005.1 | 1 | 330 |

Genomic Map