Assay Details
Target Gene Details
Entrez Gene ID: | 14538 |
Gene Name: | glucosaminyl (N-acetyl) transferase 2, I-branching enzyme |
Gene Aliases: |
5330430K10Rik, IGnT, IGnTA, IGnTB, IGnTC |
Location: |
Chr.13:40859768-40960892 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gcnt2 | NM_023887.4 | 1 | 1312 | NP_076376.3 |
| AK077598.1 | 1 | 949 | BAC36889.1 | |
| AK156328.1 | 1 | 848 | BAE33677.1 | |
| AY236874.1 | 1 | 1112 | AAO86064.1 | |
| AY435148.1 | 1 | 725 | AAR95649.1 | |
| BC094572.1 | 1 | 1312 | AAH94572.1 | |
| BQ892107.1 | 1 | 483 |
More Information
Set Membership: |
|

Genomic Map