Assay Details
Target Gene Details
Entrez Gene ID: | 14538 |
Gene Name: | glucosaminyl (N-acetyl) transferase 2, I-branching enzyme |
Gene Aliases: |
5330430K10Rik, IGnT, IGnTA, IGnTB, IGnTC |
Location: |
Chr.13:40859768-40960892 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gcnt2 | NM_008105.3 | NP_032131.2 | ||
| NM_023887.4 | NP_076376.3 | |||
| NM_133219.1 | 1 | 389 | NP_573482.1 | |
| AB037597.1 | BAB03496.1 | |||
| AK019924.1 | 1 | 374 | BAB31918.1 | |
| AK054226.1 | BAC35697.1 | |||
| AK077598.1 | BAC36889.1 | |||
| AK080061.1 | ||||
| AK156328.1 | BAE33677.1 | |||
| AY236873.1 | AAO86063.1 | |||
| AY236874.1 | AAO86064.1 | |||
| AY236875.1 | 1 | 422 | AAO86065.1 | |
| AY435148.1 | AAR95649.1 | |||
| AY435149.1 | AAR95650.1 | |||
| AY435150.1 | 1 | 140 | AAR95651.1 | |
| BC054845.1 | 1 | 371 | AAH54845.1 | |
| BC094572.1 | AAH94572.1 | |||
| BQ892107.1 | ||||
| BY208177.1 | ||||
| U68182.1 | AAB39621.1 |
More Information
Set Membership: |
|

Genomic Map