Assay Details
Target Gene Details
Entrez Gene ID: | 218506 |
Gene Name: | mitochondrial ribosomal protein S27 |
Gene Aliases: |
2610028H14Rik |
Location: |
Chr.13:99344767-99415562 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrps27 | NM_173757.3 | 11 | 1439 | NP_776118.2 |
| AK076008.1 | 11 | 1438 | BAC36116.1 | |
| AK087686.1 | 11 | 1436 | ||
| AK172922.1 | 3 | 5231 | BAD32200.1 | |
| AK185458.1 | 1 | 128 | ||
| BC049149.1 | 11 | 1366 | AAH49149.1 |
More Information
Set Membership: |
|

Genomic Map