Assay Details
Target Gene Details
Entrez Gene ID: | 22151 |
Gene Name: | tubulin, beta 2A class IIA |
Gene Aliases: |
M(beta)2, Tubb2 |
Location: |
Chr.13:34074278-34078008 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tubb2a | NM_009450.2 | 4 | 1398 | NP_033476.1 |
| AK134545.1 | 4 | 1397 | BAE22178.1 | |
| BC003475.1 | 1 | 878 | AAH03475.1 | |
| BC055441.1 | 4 | 1388 | AAH55441.1 | |
| CB522913.1 | 1 | 212 | ||
| M28739.1 | 1 | 284 | AAA40508.1 |
More Information
Set Membership: |
|

Genomic Map