Assay Details
Target Gene Details
Entrez Gene ID: | 67963 |
Gene Name: | Niemann-Pick type C2 |
Gene Aliases: |
2700012J19Rik, AA408070, AU045843, HE1 |
Location: |
Chr.12:84754559-84773112 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Npc2 | NM_023409.4 | 1 | 165 | NP_075898.1 |
| AI391386.1 | 1 | 165 | ||
| AK008603.1 | BAB25771.1 | |||
| AK009127.1 | BAB26090.1 | |||
| AK012239.1 | ||||
| AK149805.1 | ||||
| AK151067.1 | BAE30083.1 | |||
| AK151133.1 | BAE30141.1 | |||
| AK152236.1 | BAE31061.1 | |||
| AK153020.1 | BAE31654.1 | |||
| AK153203.1 | BAE31803.1 | |||
| AK154838.1 | BAE32866.1 | |||
| AK158624.1 | 1 | 133 | BAE34585.1 | |
| AK159154.1 | BAE34861.1 | |||
| AK165263.1 | BAE38111.1 | |||
| AK167869.1 | BAE39885.1 | |||
| AK170327.1 | BAE41721.1 | |||
| BC003471.1 | AAH03471.1 | |||
| BC007190.1 | AAH07190.1 |

Genomic Map