Assay Details
Target Gene Details
Entrez Gene ID: | 67963 |
Gene Name: | Niemann-Pick type C2 |
Gene Aliases: |
2700012J19Rik, AA408070, AU045843, HE1 |
Location: |
Chr.12:84754559-84773112 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Npc2 | NM_023409.4 | 4 | 2818 | NP_075898.1 |
AK154838.1 | 4 | 2605 | BAE32866.1 | |
BE952809.1 | 1 | 461 |
