Assay Details
Target Gene Details
Entrez Gene ID: | 66410 |
Gene Name: | mitochondrial transcription termination factor 3 |
Gene Aliases: |
2410017I18Rik, Mterfd1 |
Location: |
Chr.13:66905456-66933072 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mterf3 | NM_025547.3 | 2 | 422 | NP_079823.2 |
| AK002299.1 | 2 | 398 | ||
| AK010525.1 | 2 | 417 | BAB27006.1 | |
| AK011934.1 | 2 | 410 | BAB27924.3 | |
| AK034645.1 | 2 | 389 | ||
| BC025173.1 | 2 | 391 | AAH25173.1 | |
| BC085282.1 | 2 | 392 | AAH85282.1 | |
| BY269653.1 |

Genomic Map