Assay Details
Target Gene Details
Entrez Gene ID: | 17828 |
Gene Name: | biogenesis of lysosomal organelles complex-1, subunit 5, muted |
Gene Aliases: |
1810074A19Rik, Muted, mu |
Location: |
Chr.13:38602706-38635884 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bloc1s5 | NM_139063.1 | 5 | 1547 | NP_620702.1 |
AF426433.1 | 5 | 1547 | AAL99384.1 | |
AK075808.1 | 5 | 1493 | BAC35976.1 | |
AK170614.1 | 5 | 1497 | BAE41913.1 | |
AK214770.1 | ||||
BC023184.1 | 5 | 1566 | AAH23184.1 | |
BC024720.1 | 5 | 1504 | AAH24720.1 |
