Assay Details
Target Gene Details
Entrez Gene ID: | 19356 |
Gene Name: | RAD17 homolog (S. pombe) |
Gene Aliases: |
MmRad24 |
Location: |
Chr.13:100617164-100651061 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rad17 | NM_001044371.2 | 3 | 259 | NP_001037836.1 |
NM_001283011.1 | 3 | 259 | NP_001269940.1 | |
NM_011233.3 | 3 | 387 | NP_035363.2 | |
AF085737.1 | 3 | 310 | AAC36335.1 | |
AJ011923.1 | 2 | 160 | CAA09868.1 | |
AK134277.1 | 3 | 214 | BAE22079.1 | |
AK162016.1 | 3 | 246 | ||
AK167358.1 | 3 | 202 | ||
BC066855.1 | 3 | 377 | AAH66855.1 | |
BY129390.1 | 3 | 261 | ||
CF916039.1 | 3 | 317 |
