Assay Details
Target Gene Details
Entrez Gene ID: | 218397 |
Gene Name: | RAS p21 protein activator 1 |
Gene Aliases: |
Gap, RasGAP, Rasa |
Location: |
Chr.13:85214699-85289493 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rasa1 | NM_145452.3 | 25 | 3860 | NP_663427.2 |
| AK133354.1 | 21 | 2420 | BAE21614.1 | |
| AK161497.1 | 15 | 1657 | BAE36423.1 |
More Information
Set Membership: |
|

Genomic Map