Assay Details
Target Gene Details
Entrez Gene ID: | 11549 |
Gene Name: | adrenergic receptor, alpha 1a |
Gene Aliases: |
Adra1c |
Location: |
Chr.14:66635017-66771240 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adra1a | NM_001271759.1 | 1 | 234 | NP_001258688.1 |
| NM_001271760.1 | 1 | 234 | NP_001258689.1 | |
| NM_001271761.1 | 1 | 234 | NP_001258690.1 | |
| NM_013461.4 | 1 | 234 | NP_038489.2 | |
| AK042759.1 | 1 | 158 | BAC31355.1 | |
| AK050183.1 | 1 | 177 | ||
| AK079597.1 | 1 | 227 | BAC37694.1 | |
| AK085653.1 | 1 | 235 | BAC39495.1 | |
| AK138919.1 | 1 | 145 |
More Information
Set Membership: |
|

Genomic Map