Assay Details
Target Gene Details
Entrez Gene ID: | 114874 |
Gene Name: | DDHD domain containing 1 |
Gene Aliases: |
Mir5131, PA-PLA1 |
Location: |
Chr.14:45593171-45658143 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddhd1 | NM_001039106.3 | 12 | 4186 | NP_001034195.2 |
| NM_001042719.2 | 14 | 4372 | NP_001036184.1 | |
| NM_001284399.1 | 15 | 4344 | NP_001271328.1 | |
| NM_176845.5 | 13 | 4270 | NP_789815.3 | |
| AK129425.1 | 10 | 3597 | BAC98235.1 |
More Information
Set Membership: |
|

Genomic Map