Assay Details
Target Gene Details
Entrez Gene ID: | 238057 |
Gene Name: | growth differentiation factor 7 |
Gene Aliases: |
BMP12 |
Location: |
Chr.12:8291018-8302088 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gdf7 | NM_013527.1 | 2 | 557 | NP_038555.1 |
| AF525752.1 | 2 | 557 | AAP97721.1 |
Target Gene Details
Entrez Gene ID: | 68832 |
Gene Name: | lipid droplet associated hydrolase |
Gene Aliases: |
1110057K04Rik |
Location: |
Chr.12:8208100-8299566 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ldah |
More Information
Set Membership: |
|

Genomic Map