Assay Details
Target Gene Details
Entrez Gene ID: | 68054 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 12 |
Gene Aliases: |
4632419J12Rik |
Location: |
Chr.12:104028769-104044443 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpina12 | NM_026535.2 | 3 | 1011 | NP_080811.1 |
| AK014346.1 | 3 | 953 | BAB29287.1 | |
| AK014589.1 | 3 | 1011 | BAB29447.1 | |
| AY326419.1 | 2 | 844 | AAP88383.1 | |
| BC062143.1 | 3 | 965 | AAH62143.1 |
More Information
Set Membership: |
|

Genomic Map