Assay Details
Target Gene Details
Entrez Gene ID: | 28200 |
Gene Name: | dehydrogenase/reductase (SDR family) member 4 |
Gene Aliases: |
AI043103, AI790593, CR, D14Ucla2, NDRD, PHCR, PSCD, RRD, mouNRDR |
Location: |
Chr.14:55478603-55490340 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dhrs4 | NM_001037938.2 | NP_001033027.2 | ||
| NM_030686.2 | 5 | 730 | NP_109611.1 | |
| AB045132.1 | BAB18776.1 | |||
| AK002691.1 | BAB22286.1 | |||
| AK018788.1 | 5 | 613 | BAB31411.1 | |
| AK146988.1 | BAE27591.1 | |||
| AW125533.1 | 1 | 233 | ||
| BC003484.1 | AAH03484.1 | |||
| BC054361.1 | AAH54361.1 |
More Information
Set Membership: |
|

Genomic Map