Assay Details
Target Gene Details
Entrez Gene ID: | 218639 |
Gene Name: | ADP-ribosylation factor-like 15 |
Gene Aliases: |
A430036I03, Arfrp2, C230032K13Rik |
Location: |
Chr.13:113794506-114157462 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arl15 | NM_172595.4 | 5 | 2893 | NP_766183.1 |
| AK040374.1 | ||||
| AK048736.1 | 5 | 2776 | BAC33440.1 | |
| AK052164.1 | 5 | 2842 | BAC34864.1 |

Genomic Map