Assay Details
Target Gene Details
Entrez Gene ID: | 66092 |
Gene Name: | growth hormone inducible transmembrane protein |
Gene Aliases: |
1010001P14Rik, C77840, MICS1, PTD010, Tmbim5 |
Location: |
Chr.14:37120444-37135327 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ghitm | NM_001199122.1 | 9 | 2344 | NP_001186051.1 |
NM_078478.5 | 9 | 2297 | NP_510963.1 | |
AF412297.1 | 9 | 2241 | AAL07803.1 | |
AK088883.1 | 1 | 270 | ||
AK093887.1 | 9 | 2273 | BAC04243.1 | |
AK134592.1 | 9 | 2227 | BAE22197.1 | |
AK146433.1 | 9 | 2204 | BAE27167.1 | |
AK146853.1 | 9 | 2260 | BAE27483.1 | |
AK149244.1 | 1 | 791 | ||
AK158904.1 | 9 | 2287 | BAE34723.1 | |
AK168678.1 | 9 | 2345 | BAE40527.1 | |
AK168800.1 | 9 | 2253 | BAE40631.1 |
