Assay Details
Target Gene Details
Entrez Gene ID: | 14619 |
Gene Name: | gap junction protein, beta 2 |
Gene Aliases: |
AI325222, Cnx26, Cx26, Gjb-2 |
Location: |
Chr.14:57098602-57104702 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gjb2 | NM_008125.3 | 2 | 605 | NP_032151.1 |
| AK133942.1 | 2 | 520 | BAE21940.1 | |
| AK135055.1 | 2 | 606 | BAE22401.1 | |
| AK146485.1 | 2 | 606 | BAE27206.1 | |
| AK157920.1 | 2 | 606 | BAE34264.1 | |
| AK160905.1 | 2 | 606 | BAE36083.1 | |
| BC013634.1 | 2 | 580 | AAH13634.1 | |
| BC051437.1 | 2 | 550 | AAH51437.1 | |
| M63803.1 | 2 | 575 | AAA37276.1 |
More Information
Set Membership: |
|

Genomic Map