Assay Details
Target Gene Details
Entrez Gene ID: | 16413 |
Gene Name: | integrin beta 1 binding protein 1 |
Gene Aliases: |
AI449260, AU019480, Icap1 |
Location: |
Chr.12:21267246-21286279 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Itgb1bp1 | NM_008403.4 | 7 | 1310 | NP_032429.1 |
AJ001373.1 | CAA04706.1 | |||
AK075577.1 | 7 | 1311 | BAC35832.1 | |
AK089937.1 | 7 | 1424 | BAC41007.1 |
