Assay Details
Target Gene Details
Entrez Gene ID: | 97895 |
Gene Name: | NLR family, pyrin domain containing 4F |
Gene Aliases: |
C330026N02Rik, C86045, Nalp-kappa, Nalp4f |
Location: |
Chr.13:65177111-65205716 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nlrp4f | NM_175290.4 | 10 | 3068 | NP_780499.3 |
AK049352.1 | 8 | 1471 | BAC33701.1 | |
AY596201.1 | 10 | 3046 | AAU06322.1 | |
BC098479.1 | 8 | 1490 | AAH98479.1 | |
CD541102.1 |
