Assay Details
Target Gene Details
Entrez Gene ID: | 142687 |
Gene Name: | ankyrin repeat and SOCS box-containing 14 |
Gene Aliases: |
Asb-14 |
Location: |
Chr.14:26894591-26916100 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asb14 | NM_001170748.1 | 10 | 1828 | NP_001164219.1 |
| AF403042.1 | 5 | 1245 | AAL57361.1 | |
| AK164800.1 | 4 | 891 | BAE37925.1 | |
| BC145002.1 | 5 | 1214 |
More Information
Set Membership: |
|

Genomic Map