Assay Details
Target Gene Details
Entrez Gene ID: | 105841 |
Gene Name: | DENN/MADD domain containing 3 |
Gene Aliases: |
AI447457, E030003N15Rik |
Location: |
Chr.15:73512560-73572242 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dennd3 | NM_001081066.1 | 23 | 5049 | NP_001074535.1 |
| AK053114.1 | 13 | 3383 | BAC35269.1 | |
| AK173046.1 | 23 | 4949 | BAD32324.1 | |
| BC132389.1 | 23 | 4937 | ||
| BC137791.1 | 23 | 4937 |

Genomic Map