Assay Details
Target Gene Details
Entrez Gene ID: | 105847 |
Gene Name: | lipase maturation factor 2 |
Gene Aliases: |
AI451006, Tmem112b, Tmem153 |
Location: |
Chr.15:89351004-89355659 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lmf2 | NM_178919.4 | 2 | 432 | NP_849250.1 |
AK042427.1 | 2 | 391 | ||
AK084606.1 | 2 | 258 | BAC39228.1 | |
AK085435.1 | 2 | 383 | ||
AK149916.1 | 2 | 384 | BAE29164.1 | |
AK149934.1 | 2 | 433 | BAE29177.1 | |
AK158371.1 | 2 | 406 | ||
AK171488.1 | 2 | 413 | BAE42488.1 | |
AK198725.1 | 1 | 85 | ||
BC079532.1 | 2 | 328 | AAH79532.1 |
