Assay Details
Target Gene Details
Entrez Gene ID: | 20363 |
Gene Name: | selenoprotein P, plasma, 1 |
Gene Aliases: |
AU018766, D15Ucla1, Se-P, selp |
Location: |
Chr.15:3270767-3280508 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sepp1 | NM_001042613.1 | 5 | 1076 | NP_001036078.1 |
NM_001042614.1 | 5 | 1159 | NP_001036079.1 | |
NM_009155.3 | 5 | 1093 | NP_033181.3 | |
AK002450.1 | 5 | 1075 | BAC55246.3 | |
AK077265.1 | 5 | 1075 | BAC55264.3 | |
AK146774.1 | 5 | 1075 | BAE27423.1 | |
AK148623.1 | 5 | 1075 | BAE28626.1 | |
AK159269.1 | 5 | 1075 | BAE34948.1 | |
AK159524.1 | 5 | 1076 | BAE35153.1 | |
AK159894.1 | 5 | 1072 | BAE35460.1 | |
AK159911.1 | 5 | 1076 | BAE35474.1 | |
AK159935.1 | 5 | 1075 | BAE35495.1 | |
AK167611.1 | 5 | 1075 | BAE39664.1 | |
AK168463.1 | 5 | 1072 | BAE40359.1 | |
AK168583.1 | 5 | 1072 | BAE40452.1 | |
AK168631.1 | 5 | 1075 | BAE40491.1 | |
AK168747.1 | 5 | 1075 | BAE40587.1 | |
AK168926.1 | 5 | 1075 | BAE40739.1 | |
AK169011.1 | 5 | 1074 | BAE40808.1 | |
AK169158.1 | 5 | 1074 | BAE40937.1 | |
AK169362.1 | 5 | 1075 | BAE41111.1 | |
BC001991.1 | 5 | 1076 | AAH01991.2 | |
X99807.1 | 4 | 1136 |
