Assay Details
Target Gene Details
Entrez Gene ID: | 26559 |
Gene Name: | hormonally upregulated Neu-associated kinase |
Gene Aliases: |
Bstk1, Mak-v |
Location: |
Chr.16:90385797-90499555 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hunk | NM_015755.2 | 2 | 509 | NP_056570.1 |
| AF055919.2 | 2 | 509 | AAC61489.1 | |
| AF167987.1 | 2 | 521 | AAF35282.1 | |
| AK144639.1 | 2 | 492 | BAE25983.1 | |
| AK147425.1 | 2 | 894 | ||
| AK147734.1 | 2 | 263 | BAE28103.1 | |
| BC137788.1 | 2 | 570 | ||
| BC137790.1 | 2 | 569 |
More Information
Set Membership: |
|

Genomic Map